Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281293.1 | NP_001268222.1 | 2 | 2555 | ||
NM_001281294.1 | NP_001268223.1 | 3 | 2447 | |||
NM_012482.4 | NP_036614.1 | 2 | 2551 | |||
XM_017000887.1 | XP_016856376.1 | 5 | 3936 | |||
XM_017000888.1 | XP_016856377.1 | 5 | 2894 | |||
GenBank mRNA | AF125158.1 | - | 1 | 2447 | ||
AJ132591.1 | - | 1 | 1968 | |||
AJ132592.1 | - | 2 | 2531 | |||
AK021707.1 | - | 1 | 854 | |||
AK022916.1 | - | 2 | 2482 | |||
AK022921.1 | - | 1 | 1181 | |||
AK094701.1 | - | 3 | 2382 | |||
AK297233.1 | - | 3 | 2405 | |||
BC051905.1 | - | 1 | 2440 | 6165292 | ||
BC060820.1 | - | 2 | 2531 | 30344180 |