Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012329.2 | NP_036461.2 | 5 | 641 | ||
XM_006721795.2 | XP_006721858.1 | 6 | 747 | |||
GenBank mRNA | AK303810.1 | - | 5 | 486 | ||
AK312760.1 | - | 5 | 520 | |||
AY424289.1 | - | 5 | 621 | |||
BC026324.1 | - | 5 | 613 | 4794375 | ||
BI546449.1 | - | 5 | 653 | |||
CR541893.1 | - | 5 | 356 | |||
X85750.1 | - | 5 | 437 |