Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012329.2 | NP_036461.2 | 3 | 469 | ||
XM_006721795.2 | XP_006721858.1 | 3 | 482 | |||
GenBank mRNA | AK303810.1 | - | 3 | 314 | ||
AK312760.1 | - | 3 | 348 | |||
AY424289.1 | - | 3 | 449 | |||
BC026324.1 | - | 3 | 441 | 4794375 | ||
BI546449.1 | - | 3 | 481 | |||
CR541893.1 | - | 3 | 184 | |||
X85750.1 | - | 3 | 265 |