Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001204204.2 | NP_001191133.1 | 9 | 871 | ||
NM_001291932.1 | NP_001278861.1 | 10 | 1044 | |||
NM_012429.4 | NP_036561.1 | 11 | 1120 | |||
NM_033382.2 | NP_203740.1 | 11 | 1120 | |||
GenBank mRNA | AB033012.2 | - | 11 | 1019 | ||
AK056163.1 | - | 5 | 489 | |||
AK091402.1 | - | 9 | 907 | |||
AK223587.1 | - | 11 | 1080 | |||
AK296540.1 | - | 10 | 934 | |||
AK303751.1 | - | 9 | 767 | |||
AK314390.1 | - | 11 | 1053 | |||
AL096881.1 | - | 11 | 1117 | |||
AY927437.1 | - | LIT3470 AY927437 |
6 | 454 | ||
AY927438.1 | - | 9 | 742 | |||
AY927439.1 | - | 10 | 849 | |||
BC058915.1 | - | 11 | 1024 | 5178251 | ||
CR456571.1 | - | 11 | 1017 |