Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204204.2 | NP_001191133.1 | 4 | 375 | ||
NM_001291932.1 | NP_001278861.1 | 5 | 548 | |||
NM_012429.4 | NP_036561.1 | 6 | 624 | |||
NM_033382.2 | NP_203740.1 | 6 | 624 | |||
GenBank mRNA | AB033012.2 | - | 6 | 523 | ||
AK056163.1 | - | 1 | 100 | |||
AK091402.1 | - | 4 | 411 | |||
AK223587.1 | - | 6 | 584 | |||
AK296540.1 | - | 5 | 438 | |||
AK303751.1 | - | 4 | 271 | |||
AK314390.1 | - | 6 | 557 | |||
AL096881.1 | - | 6 | 621 | |||
AY927438.1 | - | 5 | 353 | |||
AY927439.1 | - | 5 | 353 | |||
BC058915.1 | - | 6 | 528 | 5178251 | ||
CR456571.1 | - | 6 | 521 | |||
DC375707.1 | - | 5 | 549 |