Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX2 SIRNA Assays ›
Gene Symbol:
RBFOX2
Gene Name:
RNA binding protein, fox-1 homolog 2
Gene Aliases:
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location:
Chr.22: 35738736 - 36028892 on Build GRCh38
UniGene ID:
Hs.282998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24048
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031695.2
NP_001026865.1
3
680
NM_001082576.1
NP_001076045.1
3
680
NM_001082577.1
NP_001076046.1
3
680
NM_001082578.1
NP_001076047.1
4
640
NM_001082579.1
NP_001076048.1
4
637
NM_014309.2
NP_055124.1
3
683
XM_005261428.2
XP_005261485.1
4
995
XM_005261429.2
XP_005261486.1
4
992
XM_005261430.2
XP_005261487.1
4
995
XM_005261431.2
XP_005261488.1
4
995
XM_005261432.2
XP_005261489.1
4
992
XM_005261433.2
XP_005261490.1
4
995
XM_005261435.1
XP_005261492.1
3
684
XM_005261437.1
XP_005261494.1
3
687
XM_006724185.2
XP_006724248.1
4
995
XM_006724186.2
XP_006724249.1
4
992
XM_006724187.2
XP_006724250.1
4
995
XM_006724188.2
XP_006724251.1
4
995
XM_006724189.3
XP_006724252.1
4
688
XM_006724190.1
XP_006724253.1
4
382
XM_006724191.1
XP_006724254.1
3
687
XM_006724192.2
XP_006724255.1
3
517
XM_006724193.3
XP_006724256.1
4
995
XM_006724194.2
XP_006724257.1
3
459
XM_017028686.1
XP_016884175.1
4
374
XM_017028687.1
XP_016884176.1
4
555
XM_017028688.1
XP_016884177.1
4
992
XM_017028689.1
XP_016884178.1
4
460
XM_017028690.1
XP_016884179.1
3
437
XM_017028691.1
XP_016884180.1
3
434
XM_017028692.1
XP_016884181.1
4
729
XM_017028693.1
XP_016884182.1
3
687
XM_017028694.1
XP_016884183.1
3
684
XM_017028695.1
XP_016884184.1
4
444
XM_017028696.1
XP_016884185.1
3
687
XM_017028697.1
XP_016884186.1
4
1464
XM_017028698.1
XP_016884187.1
3
684
GenBank mRNA
AB649123.1
-
3
315
AF229058.1
-
3
485
AK055213.1
-
4
601
AK291460.1
-
3
564
AK314632.1
-
3
499
AL009266.1
-
3
488
AM419009.1
-
3
339
AY072786.1
-
4
450
BC013115.1
-
3
504
4346741
BC025281.1
-
3
503
4555865
CR456559.1
-
3
381
DQ778625.1
-
4
528
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_053104, NM_001110828, NM_001286419, NM_001110830, NM_001110827, and others.
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
fibroblast growth factor receptor signaling pathway
regulation of definitive erythrocyte differentiation
RNA metabolic process
radial glia guided migration of Purkinje cell
intracellular estrogen receptor signaling pathway
regulation of cell proliferation
negative regulation of transcription, DNA-templated
dendrite morphogenesis
neuromuscular process controlling balance
nucleotide binding
transcription corepressor activity
RNA binding
mRNA binding
protein binding
transcription factor binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline