Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014437.2 | NP_001014437.1 | 10 | 1133 | ||
NM_001194997.1 | NP_001181926.1 | 10 | 1133 | |||
NM_001751.5 | NP_001742.1 | 9 | 884 | |||
NM_139273.3 | NP_644802.1 | 9 | 884 | |||
NR_036542.1 | 10 | 1180 | ||||
XM_006718341.3 | XP_006718404.1 | 9 | 907 | |||
XM_017018389.1 | XP_016873878.1 | 9 | 907 | |||
XM_017018390.1 | XP_016873879.1 | 10 | 1147 | |||
XM_017018391.1 | XP_016873880.1 | 7 | 1131 | |||
XM_017018392.1 | XP_016873881.1 | 9 | 1069 | |||
GenBank mRNA | AB208814.1 | - | 8 | 3058 | ||
AF288206.1 | - | 9 | 817 | |||
AF288207.1 | - | 9 | 817 | |||
AK096313.1 | - | 10 | 1147 | |||
AK222614.1 | - | 9 | 825 | |||
AK289432.1 | - | 9 | 857 | |||
AK295371.1 | - | 9 | 775 | |||
AK296763.1 | - | 10 | 998 | |||
AK298514.1 | - | 10 | 1100 | |||
AK302644.1 | - | 10 | 1100 | |||
AK308005.1 | - | 10 | 1053 | |||
BC002880.2 | - | 9 | 870 | 3940167 | ||
BT009913.1 | - | 9 | 800 | |||
BX647906.1 | - | 10 | 1107 | |||
L06845.1 | - | 7 | 671 |