Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014437.2 | NP_001014437.1 | 18 | 2108 | ||
NM_001194997.1 | NP_001181926.1 | 18 | 2108 | |||
NM_001751.5 | NP_001742.1 | 17 | 1859 | |||
NM_139273.3 | NP_644802.1 | 17 | 1859 | |||
NR_036542.1 | 18 | 2155 | ||||
XM_006718341.3 | XP_006718404.1 | 17 | 1882 | |||
XM_017018389.1 | XP_016873878.1 | 17 | 1882 | |||
XM_017018390.1 | XP_016873879.1 | 18 | 2122 | |||
XM_017018391.1 | XP_016873880.1 | 15 | 2106 | |||
XM_017018392.1 | XP_016873881.1 | 17 | 2044 | |||
GenBank mRNA | AB208814.1 | - | 14 | 5076 | ||
AF288206.1 | - | 17 | 1792 | |||
AF288207.1 | - | 17 | 1792 | |||
AK096313.1 | - | 18 | 2122 | |||
AK222614.1 | - | 17 | 1800 | |||
AK289432.1 | - | 17 | 1832 | |||
AK295371.1 | - | 17 | 1750 | |||
AK296763.1 | - | 18 | 1973 | |||
AK298514.1 | - | 18 | 2075 | |||
AK302644.1 | - | 18 | 2075 | |||
AK308005.1 | - | 18 | 2026 | |||
BC002880.2 | - | 17 | 1845 | 3940167 | ||
BT009913.1 | - | 17 | 1775 | |||
BX647906.1 | - | 18 | 2082 | |||
L06845.1 | - | 15 | 1646 |