Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014310.3 | NP_055125.2 | 2 | 338 | ||
XM_005261442.4 | XP_005261499.1 | 2 | 198 | |||
XM_011530040.2 | XP_011528342.1 | 3 | 1169 | |||
XM_017028701.1 | XP_016884190.1 | 1 | 2947 | |||
XM_017028702.1 | XP_016884191.1 | 2 | 1256 | |||
GenBank mRNA | AF279143.1 | - | 1 | 169 | ||
AL831936.1 | - | 1 | 50 | |||
BC013419.2 | - | 2 | 340 | 3604196 | ||
CR456477.1 | - | 2 | 253 |