Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012437.5 | NP_036569.1 | 3, 4 | 408 | ||
NR_052019.1 | 2, 3 | 361 | ||||
NR_052020.1 | 2, 3 | 347 | ||||
GenBank mRNA | AF086837.1 | - | 3, 4 | 318 | ||
AK024555.1 | - | 3, 4 | 385 | |||
BC000761.2 | - | 3, 4 | 337 | 2821705 | ||
BC004494.2 | - | 3, 4 | 338 | 3830132 | ||
BE278772.1 | - | 2, 3 | 262 | |||
BI117540.1 | - | 409 | ||||
BM051828.1 | - | 3, 4 | 394 | |||
BT006753.1 | - | 3, 4 | 318 | |||
CD671119.1 | - | 2, 3 | 296 |