Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001224.4 | NP_001215.1 | 10 | 1099 | ||
NM_032982.3 | NP_116764.2 | 9 | 1284 | |||
NM_032983.3 | NP_116765.2 | 8 | 1066 | |||
GenBank mRNA | AB209640.1 | - | 3 | 3956 | ||
AF314174.1 | - | 8 | 774 | |||
AF314175.1 | - | 7 | 732 | |||
AK291274.1 | - | 9 | 1076 | |||
AK303265.1 | - | 3 | 348 | |||
AK303391.1 | - | 9 | 947 | |||
BC002427.2 | - | 9 | 1056 | 3346279 | ||
BT007240.1 | - | 9 | 992 | |||
BX537669.1 | - | 1 | 1888 | |||
KU177957.1 | - | 9 | 1043 | |||
KU177958.1 | - | 7 | 793 | |||
U13021.1 | - | 9 | 1005 | |||
U13022.1 | - | 10 | 1099 |