Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PADI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012387.2 | NP_036519.2 | 15 | 1751 | ||
XM_011541150.1 | XP_011539452.1 | 13 | 1563 | |||
XM_011541151.1 | XP_011539453.1 | 13 | 1451 | |||
XM_011541152.1 | XP_011539454.1 | 12 | 1283 | |||
XM_011541157.1 | XP_011539459.1 | 12 | 1230 | |||
GenBank mRNA | AB017919.1 | - | 15 | 1749 | ||
AK290507.1 | - | 15 | 1749 | |||
AK314839.1 | - | 15 | 1751 | |||
BC025718.1 | - | 15 | 1735 | 5228777 |