Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014313.3 | NP_055128.1 | 5 | 508 | ||
XM_011541159.1 | XP_011539461.1 | 5 | 702 | |||
GenBank mRNA | AF081282.1 | - | 5 | 435 | ||
AF087880.1 | - | 5 | 499 | |||
AK311355.1 | - | 3 | 253 | |||
AK312588.1 | - | 5 | 472 | |||
AL136627.1 | - | 5 | 490 | |||
AY071927.1 | - | 5 | 485 | |||
AY358650.1 | - | 5 | 471 | |||
BC007341.2 | - | 5 | 456 | 3688672 | ||
BC066908.1 | - | 5 | 447 | 4817784 | ||
JX644905.1 | - | 5 | 508 | |||
JX644906.1 | - | 5 | 508 |