Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014313.3 | NP_055128.1 | 5, 6 | 533 | ||
XM_011541159.1 | XP_011539461.1 | 5, 6 | 727 | |||
GenBank mRNA | AF081282.1 | - | 5, 6 | 460 | ||
AF087880.1 | - | 5, 6 | 524 | |||
AK311355.1 | - | 3, 4 | 278 | |||
AK312588.1 | - | 5, 6 | 497 | |||
AL136627.1 | - | 5, 6 | 515 | |||
AY071927.1 | - | 5, 6 | 510 | |||
AY358650.1 | - | 5, 6 | 496 | |||
BC007341.2 | - | 5, 6 | 481 | 3688672 | ||
BC066908.1 | - | 5, 6 | 472 | 4817784 | ||
JX644905.1 | - | 5, 6 | 533 | |||
JX644906.1 | - | 5, 6 | 533 |