Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014315.2 | NP_055130.1 | 9 | 1265 | ||
XM_006720094.3 | XP_006720157.1 | 9 | 1745 | |||
XM_011536610.1 | XP_011534912.1 | 9 | 998 | |||
GenBank mRNA | AF113131.1 | - | 9 | 1123 | ||
AF244137.1 | - | 9 | 1200 | |||
AK001771.1 | - | 9 | 1181 | |||
AK056298.1 | - | 9 | 1726 | |||
BC002335.2 | - | 9 | 1192 | 3506591 | ||
BX161429.1 | - | 8 | 1232 | |||
CR457205.1 | - | 9 | 865 |