Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321978.1 | NP_001308907.1 | 10 | 1264 | ||
NM_001321979.1 | NP_001308908.1 | 12 | 1538 | |||
NM_014317.4 | NP_055132.2 | 12 | 1455 | |||
XM_011519437.2 | XP_011517739.1 | 9 | 1855 | |||
XM_017016011.1 | XP_016871500.1 | 10 | 933 | |||
GenBank mRNA | AB209763.1 | - | 11 | 3497 | ||
AB210838.1 | - | 12 | 1213 | |||
AF118395.1 | - | 11 | 1075 | |||
AK024802.1 | - | 12 | 1220 | |||
AK223414.1 | - | 12 | 1220 | |||
AK296288.1 | - | 5 | 525 | |||
BC049211.1 | - | 12 | 1165 | |||
BC063635.1 | - | 10 | 992 | 4397753 |