Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014322.2 | NP_055137.2 | 4 | 1438 | ||
GenBank mRNA | AF140242.1 | - | 4 | 1361 | ||
AF303588.1 | - | 4 | 1395 | |||
AK310307.1 | - | 4 | 921 | |||
BC036773.1 | - | 4 | 1390 | 5723543 | ||
BC051900.1 | - | 3 | 992 | |||
BC062622.1 | - | 4 | 1219 | |||
BC073872.1 | - | 3 | 1043 |
Set Membership: |
G-protein coupled receptor |