Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033583.2 | NP_001028755.2 | 10, 11 | 970 | ||
NM_001037171.1 | NP_001032248.1 | 11, 12 | 997 | |||
XM_005274472.2 | XP_005274529.1 | 9, 10 | 869 | |||
GenBank mRNA | AF132950.1 | - | 9, 10 | 872 | ||
AF241787.1 | - | 1, 2 | 389 | |||
AK000659.1 | - | 10, 11 | 956 | |||
AK024337.1 | - | 12, 13 | 2062 | |||
BC136671.1 | - | 10, 11 | 921 | 9020661 | ||
BC144360.1 | - | 10, 11 | 921 | 9052881 | ||
BC144370.1 | - | 9, 10 | 847 | |||
BX648512.1 | - | 11, 12 | 981 |