Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014323.2 | NP_055138.2 | 1 | 1175 | ||
NM_032050.1 | NP_114439.1 | 1 | 1175 | |||
NM_032051.1 | NP_114440.1 | 1 | 1175 | |||
NM_032052.1 | NP_114441.1 | 1 | 1175 | |||
GenBank mRNA | AB208787.1 | - | 2 | 545 | ||
AF119256.1 | - | 1 | 1174 | |||
AF242522.1 | - | 1 | 531 | |||
AF254082.1 | - | 1 | 1144 | |||
AF254083.1 | - | 1 | 1144 | |||
AF254084.1 | - | 1 | 1144 | |||
AF254085.1 | - | 1 | 1144 | |||
AK291803.1 | - | 1 | 688 | |||
AL096880.1 | - | 1 | 509 | |||
BC021091.1 | - | 1 | 722 | 4857537 |