Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167595.1 | NP_001161067.1 | 4 | 720 | ||
NM_014324.5 | NP_055139.4 | 4 | 720 | |||
NM_203382.2 | NP_976316.1 | 3 | 559 | |||
GenBank mRNA | AF047020.1 | - | 4 | 712 | ||
AF158378.1 | - | 4 | 689 | |||
AJ130733.1 | - | 4 | 712 | |||
AK000912.1 | - | 4 | 705 | |||
AK022765.1 | - | 4 | 690 | |||
AK222831.1 | - | 4 | 692 | |||
AK225749.1 | - | 4 | 729 | |||
AK292988.1 | - | 3 | 509 | |||
AL545355.3 | - | 4 | 633 | |||
AY330488.1 | - | 3 | 472 | |||
AY330489.1 | - | 4 | 633 | |||
AY935981.1 | - | 4 | 633 | |||
BC009471.1 | - | 3 | 475 | 2958112 | ||
BG708726.1 | - | 3 | 495 | |||
BQ941482.1 | - | 2 | 229 | |||
BT007193.1 | - | 3 | 463 | |||
EF560721.1 | - | 4 | 643 | |||
FJ498906.1 | - | 3 | 475 | |||
FJ498907.1 | - | 3 | 475 | |||
FJ498908.1 | - | 3 | 475 |
Gene Symbol: C1QTNF3-AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037951.1 | 8 | 980 | |||
GenBank mRNA | AB209261.1 | - | 8 | 989 |