Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105237.2 | NP_001098707.1 | 10 | 1394 | ||
NM_001276471.1 | NP_001263400.1 | 10 | 1412 | |||
NM_014325.3 | NP_055140.1 | 10 | 1313 | |||
XM_011538125.1 | XP_011536427.1 | 6 | 637 | |||
XM_017019121.1 | XP_016874610.1 | 11 | 2142 | |||
XM_017019122.1 | XP_016874611.1 | 10 | 1719 | |||
XM_017019123.1 | XP_016874612.1 | 11 | 1772 | |||
GenBank mRNA | AB030656.1 | - | 10 | 1282 | ||
AB209909.1 | - | 10 | 1268 | |||
AK023205.1 | - | 10 | 1283 | |||
AK096363.1 | - | 10 | 1412 | |||
AK223073.1 | - | 10 | 1281 | |||
AK297463.1 | - | 10 | 1146 | |||
AK304839.1 | - | 10 | 1049 | |||
AK311315.1 | - | 4 | 238 | |||
AK316065.1 | - | 10 | 1109 | |||
AK316504.1 | - | 9 | 1034 | |||
AL162070.1 | - | 10 | 1251 | |||
AM849477.1 | - | 10 | 1287 | |||
AM849478.1 | - | 10 | 1394 | |||
BC002342.2 | - | 10 | 1267 | 2822459 |