Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145125.1 | NP_001138597.1 | 3 | 730 | ||
NM_001291663.1 | NP_001278592.1 | 5 | 892 | |||
NM_013446.3 | NP_038474.2 | 3 | 730 | |||
XM_011515996.1 | XP_011514298.1 | 2 | 519 | |||
XM_011515997.2 | XP_011514299.1 | 3 | 787 | |||
XM_011515998.1 | XP_011514300.1 | 3 | 472 | |||
GenBank mRNA | AB209298.1 | - | 3 | 497 | ||
AF117233.1 | - | 3 | 660 | |||
AF192784.1 | - | 3 | 625 | |||
AK027137.1 | - | 2 | 242 | |||
AK127030.1 | - | 4 | 1347 | |||
AK315552.1 | - | 3 | 529 | |||
AL136812.1 | - | 3 | 628 | |||
AM236048.1 | - | 3 | 511 | |||
BC025955.1 | - | 3 | 663 | 4869484 | ||
BC037400.1 | - | 3 | 515 | 5176176 | ||
BC064838.1 | - | 3 | 508 | 6173715 | ||
BC127216.1 | - | 1 | 129 |