Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24202
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
12
1714
NM_001281772.2
NP_001268701.1
14
1913
NM_001281773.2
NP_001268702.1
13
1860
NM_001281774.2
NP_001268703.1
13
1860
NM_001281775.2
NP_001268704.1
12
1789
NM_001281776.2
NP_001268705.1
12
1789
NM_001281777.2
NP_001268706.1
12
1714
NM_001281778.2
NP_001268707.1
12
1714
NM_001281779.2
NP_001268708.1
13
1920
NM_001281780.2
NP_001268709.1
13
1920
NM_001281782.2
NP_001268711.1
12
1714
NM_001281783.2
NP_001268712.1
12
1789
NM_012408.5
NP_036540.3
12
1789
NM_183047.3
NP_898868.1
12
1789
NM_183048.3
NP_898869.1
12
1714
XM_005260356.4
XP_005260413.1
12
1826
XM_005260358.2
XP_005260415.1
11
1568
XM_005260360.4
XP_005260417.1
12
2291
XM_005260362.2
XP_005260419.1
12
1566
XM_005260364.2
XP_005260421.1
12
1551
XM_005260366.2
XP_005260423.1
12
1582
XM_006723760.3
XP_006723823.1
12
1964
XM_006723762.3
XP_006723825.1
12
1665
XM_006723763.1
XP_006723826.1
12
1580
XM_006723766.1
XP_006723829.1
12
1524
XM_011528749.2
XP_011527051.1
12
2470
XM_011528751.1
XP_011527053.1
12
1574
XM_011528753.1
XP_011527055.1
13
1585
XM_011528754.1
XP_011527056.1
12
1512
XM_017027760.1
XP_016883249.1
11
1571
XM_017027761.1
XP_016883250.1
12
2037
XM_017027762.1
XP_016883251.1
14
2804
XM_017027763.1
XP_016883252.1
12
1717
XM_017027764.1
XP_016883253.1
12
2236
XM_017027765.1
XP_016883254.1
12
1566
XM_017027766.1
XP_016883255.1
12
1566
XM_017027767.1
XP_016883256.1
12
2381
XM_017027768.1
XP_016883257.1
12
1641
XM_017027769.1
XP_016883258.1
12
1551
XM_017027770.1
XP_016883259.1
13
1585
XM_017027771.1
XP_016883260.1
12
1551
XM_017027772.1
XP_016883261.1
12
1524
XM_017027773.1
XP_016883262.1
12
1566
XM_017027774.1
XP_016883263.1
11
1544
XM_017027775.1
XP_016883264.1
12
1512
XM_017027777.1
XP_016883266.1
11
1544
XM_017027778.1
XP_016883267.1
11
1556
XM_017027779.1
XP_016883268.1
11
1556
XM_017027780.1
XP_016883269.1
11
1571
GenBank mRNA
AB032951.1
-
12
1764
AF273045.1
-
2
387
AF454056.1
-
12
1606
AK000275.1
-
12
1531
AK026151.1
-
2
334
AK074262.1
-
2
324
AK122966.1
-
11
1410
AK225857.1
-
12
1597
AK294511.1
-
14
1723
AK295747.1
-
12
1673
AK299899.1
-
13
1672
AK309228.1
-
12
1583
AL137703.1
-
11
1525
BC030721.1
-
9
1289
BC136608.1
-
12
1545
9020598
BC144289.1
-
12
1545
9052809
BC146802.1
-
12
1555
8860316
BX641005.1
-
13
1805
DQ082998.1
-
11
1570
DQ082999.1
-
11
1570
DQ368669.1
-
11
1570
DQ368670.1
-
11
1570
DQ368671.1
-
11
1495
DQ368672.1
-
11
1495
DQ368673.1
-
11
1570
DQ368674.1
-
11
1495
DQ368675.1
-
12
1701
DQ368676.1
-
12
1701
DQ368678.1
-
11
1495
DQ368679.1
-
11
1570
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0