Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24204
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
12
1710
NM_001281772.2
NP_001268701.1
14
1909
NM_001281773.2
NP_001268702.1
13
1856
NM_001281774.2
NP_001268703.1
13
1856
NM_001281775.2
NP_001268704.1
12
1785
NM_001281776.2
NP_001268705.1
12
1785
NM_001281777.2
NP_001268706.1
12
1710
NM_001281778.2
NP_001268707.1
12
1710
NM_001281779.2
NP_001268708.1
13
1916
NM_001281780.2
NP_001268709.1
13
1916
NM_001281782.2
NP_001268711.1
12
1710
NM_001281783.2
NP_001268712.1
12
1785
NM_012408.5
NP_036540.3
12
1785
NM_183047.3
NP_898868.1
12
1785
NM_183048.3
NP_898869.1
12
1710
XM_005260356.4
XP_005260413.1
12
1822
XM_005260358.2
XP_005260415.1
11
1564
XM_005260360.4
XP_005260417.1
12
2287
XM_005260362.2
XP_005260419.1
12
1562
XM_005260364.2
XP_005260421.1
12
1547
XM_005260366.2
XP_005260423.1
12
1578
XM_006723760.3
XP_006723823.1
12
1960
XM_006723762.3
XP_006723825.1
12
1661
XM_006723763.1
XP_006723826.1
12
1576
XM_006723766.1
XP_006723829.1
12
1520
XM_011528749.2
XP_011527051.1
12
2466
XM_011528751.1
XP_011527053.1
12
1570
XM_011528753.1
XP_011527055.1
13
1581
XM_011528754.1
XP_011527056.1
12
1508
XM_017027760.1
XP_016883249.1
11
1567
XM_017027761.1
XP_016883250.1
12
2033
XM_017027762.1
XP_016883251.1
14
2800
XM_017027763.1
XP_016883252.1
12
1713
XM_017027764.1
XP_016883253.1
12
2232
XM_017027765.1
XP_016883254.1
12
1562
XM_017027766.1
XP_016883255.1
12
1562
XM_017027767.1
XP_016883256.1
12
2377
XM_017027768.1
XP_016883257.1
12
1637
XM_017027769.1
XP_016883258.1
12
1547
XM_017027770.1
XP_016883259.1
13
1581
XM_017027771.1
XP_016883260.1
12
1547
XM_017027772.1
XP_016883261.1
12
1520
XM_017027773.1
XP_016883262.1
12
1562
XM_017027774.1
XP_016883263.1
11
1540
XM_017027775.1
XP_016883264.1
12
1508
XM_017027777.1
XP_016883266.1
11
1540
XM_017027778.1
XP_016883267.1
11
1552
XM_017027779.1
XP_016883268.1
11
1552
XM_017027780.1
XP_016883269.1
11
1567
GenBank mRNA
AB032951.1
-
12
1760
AF273045.1
-
2
383
AF454056.1
-
12
1602
AK000275.1
-
12
1527
AK026151.1
-
2
330
AK074262.1
-
2
320
AK122966.1
-
11
1406
AK225857.1
-
12
1593
AK294511.1
-
14
1719
AK295747.1
-
12
1669
AK299899.1
-
13
1668
AK309228.1
-
12
1579
AL137703.1
-
11
1521
BC030721.1
-
9
1285
BC136608.1
-
12
1541
9020598
BC144289.1
-
12
1541
9052809
BC146802.1
-
12
1551
8860316
BX641005.1
-
13
1801
DQ082998.1
-
11
1566
DQ082999.1
-
11
1566
DQ368669.1
-
11
1566
DQ368670.1
-
11
1566
DQ368671.1
-
11
1491
DQ368672.1
-
11
1491
DQ368673.1
-
11
1566
DQ368674.1
-
11
1491
DQ368675.1
-
12
1697
DQ368676.1
-
12
1697
DQ368678.1
-
11
1491
DQ368679.1
-
11
1566
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0