Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207048.1 | NP_001193977.1 | 6 | 904 | ||
NM_001207049.1 | NP_001193978.1 | 6 | 829 | |||
NM_012104.4 | NP_036236.1 | 7 | 1542 | |||
NM_138971.3 | NP_620427.1 | 7 | 1410 | |||
NM_138972.3 | NP_620428.1 | 7 | 1467 | |||
NM_138973.3 | NP_620429.1 | 7 | 1335 | |||
GenBank mRNA | AB032975.2 | - | 7 | 1524 | ||
AB050436.1 | - | 7 | 1009 | |||
AB050437.1 | - | 7 | 952 | |||
AB050438.1 | - | 7 | 877 | |||
AB177847.1 | - | 6 | 766 | |||
AF161367.1 | - | 3 | 370 | |||
AF190725.1 | - | 7 | 1534 | |||
AF200193.1 | - | 7 | 1042 | |||
AF200343.1 | - | 7 | 1081 | |||
AF201468.1 | - | 7 | 1535 | |||
AF204943.1 | - | 7 | 1081 | |||
AF338816.1 | - | 7 | 1006 | |||
AF338817.1 | - | 7 | 949 | |||
AK075049.1 | - | 7 | 1017 | |||
AK296015.1 | - | 6 | 732 | |||
AK296535.1 | - | 6 | 817 | |||
BC036084.1 | - | 7 | 1537 | 5311572 | ||
BC065492.1 | - | 7 | 1542 | 6538154 |