Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012409.3 | NP_036541.2 | 2 | 323 | ||
GenBank mRNA | AF187843.1 | - | 2 | 257 | ||
AF187844.1 | - | 1 | 208 | |||
AK313404.1 | - | 2 | 324 | |||
AY358985.1 | - | 2 | 252 | |||
BC043644.2 | - | 2 | 267 | 5272576 |