Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012316.4 | NP_036448.1 | 7 | 726 | ||
XM_005270711.3 | XP_005270768.1 | 7 | 1087 | |||
GenBank mRNA | AB621806.1 | - | 7 | 715 | ||
AF060543.1 | - | 7 | 693 | |||
AK002111.1 | - | 7 | 713 | |||
AK128609.1 | - | 7 | 689 | |||
AK295615.1 | - | 7 | 841 | |||
AK301714.1 | - | 7 | 680 | |||
AK315490.1 | - | 7 | 713 | |||
BC020520.1 | - | 7 | 701 | 3913864 | ||
BT009843.1 | - | 6 | 638 |