Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012317.3 | NP_036449.1 | 1 | 422 | ||
GenBank mRNA | AB019527.1 | - | 1 | 333 | ||
BC003104.1 | - | 1 | 307 | 3503444 | ||
CR457088.1 | - | 1 | 229 |