Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014329.4 | NP_055144.3 | 9 | 1300 | ||
GenBank mRNA | AK128582.1 | - | 8 | 1774 | ||
AK291049.1 | - | 9 | 1068 | |||
AK302563.1 | - | 9 | 1066 | |||
BC043616.1 | - | 9 | 1203 | 6065418 | ||
BC053598.1 | - | 4 | 440 | |||
BC064567.1 | - | 9 | 1197 | 5575195 | ||
L26339.1 | - | 9 | 1197 |