Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031696.3 | NP_001026866.1 | 9 | 1239 | ||
NM_001291311.1 | NP_001278240.1 | 9 | 1173 | |||
NM_012268.3 | NP_036400.2 | 9 | 1463 | |||
XM_005258704.1 | XP_005258761.1 | 9 | 1384 | |||
XM_005258707.4 | XP_005258764.1 | 9 | 1166 | |||
XM_005258708.3 | XP_005258765.1 | 8 | 1059 | |||
XM_005258709.4 | XP_005258766.1 | 8 | 1047 | |||
XM_005258710.4 | XP_005258767.1 | 8 | 1088 | |||
XM_006723122.1 | XP_006723185.1 | 9 | 1551 | |||
XM_011526692.1 | XP_011524994.1 | 9 | 1546 | |||
XM_011526693.1 | XP_011524995.1 | 9 | 1266 | |||
XM_017026546.1 | XP_016882035.1 | 10 | 1482 | |||
XM_017026547.1 | XP_016882036.1 | 9 | 1050 | |||
XM_017026548.1 | XP_016882037.1 | 9 | 1201 | |||
XM_017026549.1 | XP_016882038.1 | 9 | 1304 | |||
GenBank mRNA | AK293690.1 | - | 9 | 907 | ||
BC000553.1 | - | 5 | 709 | |||
BC036327.1 | - | 9 | 964 | 5189261 | ||
BC096820.1 | - | 9 | 1162 | 30558682 | ||
U60644.1 | - | 9 | 1169 |