Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 22 | 3783 | ||
XM_005261909.1 | XP_005261966.1 | 23 | 3831 | |||
XM_005261910.2 | XP_005261967.1 | 23 | 3842 | |||
XM_005261911.4 | XP_005261968.1 | 22 | 3781 | |||
XM_006724413.2 | XP_006724476.1 | 24 | 3901 | |||
XM_011530682.2 | XP_011528984.1 | 23 | 3982 | |||
XM_017028703.1 | XP_016884192.1 | 24 | 3882 | |||
XM_017028704.1 | XP_016884193.1 | 23 | 3835 | |||
GenBank mRNA | AB002313.1 | - | 21 | 3700 | ||
AB075511.1 | - | 7 | 893 | |||
AK025701.1 | - | 9 | 1151 | |||
AK056543.1 | - | 5 | 952 | |||
AK074932.1 | - | 4 | 419 | |||
AK126394.1 | - | 3 | 492 |