Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001229.4 | NP_001220.2 | 2 | 606 | ||
NM_001278054.1 | NP_001264983.1 | 2 | 606 | |||
NM_032996.3 | NP_127463.2 | 2 | 437 | |||
NR_102732.1 | 2 | 606 | ||||
XM_005246014.2 | XP_005246071.1 | 2 | 290 | |||
XM_011542273.2 | XP_011540575.1 | 2 | 400 | |||
GenBank mRNA | AB015653.1 | - | 2 | 382 | ||
AB020979.1 | - | 2 | 361 | |||
AB209147.1 | - | 2 | 606 | |||
AF093130.1 | - | 2 | 361 | |||
AF110376.1 | - | 2 | 361 | |||
AK292080.1 | - | 2 | 378 | |||
AK292111.1 | - | 2 | 382 | |||
AK303743.1 | - | 2 | 373 | |||
AY732490.2 | - | 2 | 361 | |||
BC002452.2 | - | 2 | 367 | 3343958 | ||
BC006463.1 | - | 2 | 456 | 3542075 | ||
BT006911.1 | - | 2 | 361 | |||
DA078665.1 | - | 2 | 338 | |||
HY027003.1 | - | 2 | 400 | |||
U56390.1 | - | 2 | 364 | |||
U60521.1 | - | 2 | 406 |