Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014331.3 | NP_055146.1 | 10 | 1505 | ||
XM_011531802.2 | XP_011530104.1 | 10 | 1574 | |||
GenBank mRNA | AB026891.1 | - | 10 | 1460 | ||
AB040875.1 | - | 10 | 1360 | |||
AF200708.1 | - | 10 | 1470 | |||
AF252872.1 | - | 10 | 1456 | |||
AJ277882.1 | - | 10 | 1225 | |||
AK290359.1 | - | 10 | 1505 | |||
AK314855.1 | - | 10 | 1399 | |||
BC012087.1 | - | 10 | 1385 | 4562994 |