Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012320.3 | NP_036452.1 | 6 | 991 | ||
XM_005255886.3 | XP_005255943.1 | 6 | 839 | |||
XM_011522979.2 | XP_011521281.1 | 7 | 1060 | |||
XM_011522980.2 | XP_011521282.2 | 7 | 931 | |||
GenBank mRNA | AB017494.1 | - | 6 | 940 | ||
AK001705.1 | - | 6 | 952 | |||
AK074828.1 | - | 6 | 991 | |||
AK127898.1 | - | 2 | 1391 | |||
AK222790.1 | - | 6 | 950 | |||
AK296263.1 | - | 6 | 837 | |||
AK298494.1 | - | 2 | 362 | |||
AK300596.1 | - | 5 | 749 | |||
AL110209.1 | - | 6 | 972 | |||
AL389957.1 | - | 2 | 248 | |||
AY358425.1 | - | 6 | 928 | |||
BC011640.2 | - | 3 | 505 | |||
BC062605.1 | - | 6 | 955 | 5162981 |