Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014521.2 | NP_055336.1 | 3 | 495 | ||
XM_011510891.2 | XP_011509193.1 | 3 | 515 | |||
XM_011510892.1 | XP_011509194.1 | 3 | 504 | |||
XM_011510893.1 | XP_011509195.1 | 3 | 429 | |||
XM_011510894.2 | XP_011509196.1 | 3 | 2928 | |||
GenBank mRNA | AF147747.1 | - | 3 | 437 | ||
AK291209.1 | - | 3 | 462 | |||
BC057396.1 | - | 3 | 506 | 6138465 | ||
BX647198.1 | - | 3 | 422 | |||
BX648877.1 | - | 3 | 610 | |||
DQ232895.1 | - | 3 | 469 |