Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033578.2 | NP_001028750.1 | 5 | 817 | ||
NM_013257.4 | NP_037389.4 | 5 | 540 | |||
NM_170709.2 | NP_733827.2 | 5 | 540 | |||
GenBank mRNA | AF085233.2 | - | 4 | 331 | ||
AF169035.1 | - | 4 | 301 | |||
AK074666.1 | - | 5 | 494 | |||
AK223523.1 | - | 5 | 509 | |||
AK291428.1 | - | 5 | 817 | |||
BC015326.1 | - | 4 | 334 | 4391699 | ||
BI547053.1 | - | 6 | 579 | |||
CR536581.1 | - | 2 | 79 | |||
CR933673.1 | - | 5 | 474 |
Gene Symbol: C8orf44-SGK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204173.1 | NP_001191102.1 | 7 | 911 |