Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206524.1 | NP_001193453.1 | 7 | 1466 | ||
NM_001206542.1 | NP_001193471.1 | 7 | 1538 | |||
NM_001230.4 | NP_001221.2 | 7 | 1538 | |||
NM_032974.4 | NP_116756.2 | 9 | 1667 | |||
NM_032976.3 | NP_116758.1 | 8 | 1575 | |||
NM_032977.3 | NP_116759.2 | 9 | 1667 | |||
XM_005246907.2 | XP_005246964.1 | 9 | 1450 | |||
XM_006712796.3 | XP_006712859.1 | 8 | 1585 | |||
GenBank mRNA | AF111344.1 | - | 7 | 1157 | ||
AF111345.1 | - | 8 | 1249 | |||
AJ487678.1 | - | 7 | 1172 | |||
AJ487679.1 | - | 7 | 1241 | |||
BC042844.1 | - | 9 | 1403 | 5176698 | ||
BX089180.1 | - | 1 | 86 | |||
KU177959.1 | - | 8 | 1249 | |||
KU177960.1 | - | 6 | 1120 | |||
U60519.1 | - | 7 | 1267 | |||
U86214.1 | - | 9 | 1443 |