Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRRS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014334.2 | NP_055149.2 | 3 | 600 | ||
XM_011518453.2 | XP_011516755.1 | 3 | 413 | |||
XM_011518454.2 | XP_011516756.1 | 4 | 984 | |||
GenBank mRNA | AF155065.1 | - | 3 | 600 | ||
BF514667.1 | - | 3 | 441 |