Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRRS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014334.2 | NP_055149.2 | 4 | 824 | ||
XM_011518453.2 | XP_011516755.1 | 4 | 637 | |||
XM_011518454.2 | XP_011516756.1 | 5 | 1208 | |||
GenBank mRNA | AF155065.1 | - | 4 | 824 |