Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289905.1 | NP_001276834.1 | 4 | 529 | ||
NM_014339.6 | NP_055154.3 | 4 | 529 | |||
GenBank mRNA | BC011624.2 | - | 4 | 473 | 4053746 | |
BX334547.2 | - | 4 | 532 | |||
BX410574.2 | - | 4 | 445 | |||
CB044543.1 | - | 3 | 512 | |||
CT841520.1 | - | 4 | 452 | |||
JF305973.1 | - | 4 | 396 | |||
U58917.1 | - | 4 | 428 |