Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013281.3 | NP_037413.1 | 2 | 1853 | ||
NM_198391.2 | NP_938205.1 | 3 | 2047 | |||
XM_005260682.4 | XP_005260739.1 | 3 | 2565 | |||
XM_011529204.2 | XP_011527506.1 | 4 | 2702 | |||
XM_011529205.2 | XP_011527507.1 | 4 | 2364 | |||
GenBank mRNA | AB040902.1 | - | 2 | 1741 | ||
AF169677.1 | - | 2 | 1750 | |||
AK027297.1 | - | 2 | 1764 | |||
AK027670.1 | - | 1 | 961 | |||
AK027694.1 | - | 1 | 1482 | |||
AK074883.1 | - | 2 | 1794 | |||
AK074909.1 | - | 2 | 1722 | |||
AY358319.1 | - | 1 | 1587 | |||
BC020870.1 | - | 2 | 1760 | 4606852 |