Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308373.1 | NP_001295302.1 | 6 | 989 | ||
NM_012181.4 | NP_036313.3 | 6 | 992 | |||
XM_011527863.1 | XP_011526165.1 | 6 | 1050 | |||
XM_011527864.1 | XP_011526166.1 | 5 | 881 | |||
GenBank mRNA | AK026952.1 | - | 1580 | |||
AK313363.1 | - | 6 | 923 | |||
AY225339.1 | - | 5 | 853 | |||
AY278607.1 | - | 6 | 916 | |||
BC009966.1 | - | 5 | 696 | 4299915 | ||
BX538124.1 | - | 6 | 970 | |||
BX647405.1 | - | 6 | 969 | |||
BX647720.1 | - | 6 | 927 | |||
GQ372970.1 | - | 3 | 489 |