Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001232.3 | NP_001223.2 | 6 | 905 | ||
GenBank mRNA | AK129891.1 | - | 6 | 880 | ||
AK225959.1 | - | 6 | 767 | |||
AK293595.1 | - | 6 | 914 | |||
AK295502.1 | - | 4 | 601 | |||
AK313041.1 | - | 6 | 851 | |||
BC022288.1 | - | 6 | 814 | 4289141 | ||
D55655.1 | - | 6 | 750 |