Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193268.1 | NP_001180197.1 | 17 | 2060 | ||
NM_001193269.1 | NP_001180198.1 | 17 | 1964 | |||
NM_012155.2 | NP_036287.1 | 14 | 1425 | |||
NR_034098.1 | 12 | 1145 | ||||
XM_006723125.2 | XP_006723188.1 | 17 | 2141 | |||
XM_006723126.2 | XP_006723189.1 | 17 | 1949 | |||
XM_011526694.2 | XP_011524996.1 | 16 | 2048 | |||
XM_011526695.2 | XP_011524997.1 | 16 | 2009 | |||
XM_011526698.2 | XP_011525000.1 | 14 | 1467 | |||
XM_011526700.2 | XP_011525002.1 | 14 | 1413 | |||
XM_011526701.2 | XP_011525003.1 | 9 | 1016 | |||
XM_017026550.1 | XP_016882039.1 | 13 | 1309 | |||
GenBank mRNA | AB209773.1 | - | 11 | 1161 | ||
AF103939.1 | - | 14 | 1419 | |||
AK026616.1 | - | 12 | 1123 | |||
AK225043.1 | - | 14 | 1411 | |||
AK294668.1 | - | 17 | 1882 | |||
AK295905.1 | - | 17 | 1964 | |||
AK296258.1 | - | 17 | 1893 | |||
AK302961.1 | - | 18 | 2146 | |||
AL096717.1 | - | 14 | 1396 | |||
BC029304.1 | - | 17 | 2127 | |||
BC032630.1 | - | 13 | 1342 | 5402615 |