Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015368.3 | NP_056183.2 | 4 | 1076 | ||
XM_011542734.2 | XP_011541036.1 | 5 | 686 | |||
XM_017017464.1 | XP_016872953.1 | 5 | 1021 | |||
GenBank mRNA | AF398509.1 | - | 4 | 1074 | ||
AK074897.1 | - | 4 | 1021 | |||
AK313074.1 | - | 4 | 1046 | |||
AY359023.1 | - | 4 | 837 | |||
BC016931.1 | - | 4 | 1071 | 4390851 | ||
FM201789.1 | - | 4 | 691 |