Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012435.2 | NP_036567.2 | 11 | 1501 | ||
XM_011527893.2 | XP_011526195.1 | 11 | 1516 | |||
XM_011527894.2 | XP_011526196.1 | 10 | 1381 | |||
XM_011527895.2 | XP_011526197.1 | 10 | 1381 | |||
XM_011527896.2 | XP_011526198.1 | 13 | 1561 | |||
XM_017026567.1 | XP_016882056.1 | 12 | 1649 | |||
GenBank mRNA | AB001451.1 | - | 11 | 1375 | ||
AL360254.1 | - | 9 | 931 | |||
BC048290.1 | - | 1 | 203 | |||
BY796046.2 | - | 2 | 318 | |||
CF457169.1 | - | 2 | 303 |