Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031698.2 | NP_001026868.2 | 18 | 1874 | ||
NM_012272.2 | NP_036404.1 | 17 | 2164 | |||
XM_006719324.3 | XP_006719387.1 | 17 | 2928 | |||
XM_006719325.3 | XP_006719388.1 | 17 | 2928 | |||
XM_011538135.2 | XP_011536437.1 | 17 | 2928 | |||
XM_011538136.2 | XP_011536438.1 | 17 | 2928 | |||
XM_011538137.2 | XP_011536439.1 | 16 | 2847 | |||
XM_011538138.2 | XP_011536440.1 | 16 | 2817 | |||
XM_011538139.2 | XP_011536441.1 | 18 | 1868 | |||
XM_011538140.2 | XP_011536442.1 | 18 | 1868 | |||
XM_011538141.1 | XP_011536443.1 | 17 | 1694 | |||
XM_011538143.2 | XP_011536445.1 | 17 | 1710 | |||
XM_011538144.2 | XP_011536446.1 | 16 | 1583 | |||
XM_017019135.1 | XP_016874624.1 | 17 | 1631 | |||
XM_017019136.1 | XP_016874625.1 | 16 | 1583 | |||
XM_017019137.1 | XP_016874626.1 | 16 | 1583 | |||
XM_017019138.1 | XP_016874627.1 | 17 | 2928 | |||
XM_017019139.1 | XP_016874628.1 | 13 | 1594 | |||
GenBank mRNA | AK027117.1 | - | 3 | 155 | ||
AL137459.1 | - | 4 | 377 | |||
AL834216.1 | - | 17 | 2164 | |||
BC031917.1 | - | 15 | 1470 | |||
BC050398.2 | - | 18 | 1710 | |||
BC067364.1 | - | 17 | 1695 | 6644672 |