Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284304.1 | NP_001271233.1 | 5, 6 | 942 | ||
NM_001284305.1 | NP_001271234.1 | 4, 5 | 882 | |||
NM_014346.3 | NP_055161.1 | 4, 5 | 805 | |||
NR_104292.1 | 5, 6 | 1248 | ||||
XM_005261496.1 | XP_005261553.1 | 4, 5 | 801 | |||
XM_011530099.1 | XP_011528401.1 | 4, 5 | 801 | |||
XM_011530100.2 | XP_011528402.1 | 4, 5 | 801 | |||
XM_011530101.2 | XP_011528403.1 | 4, 5 | 801 | |||
XM_011530102.2 | XP_011528404.1 | 4, 5 | 801 | |||
XM_017028742.1 | XP_016884231.1 | 4, 5 | 801 | |||
XM_017028743.1 | XP_016884232.1 | 4, 5 | 801 | |||
XM_017028744.1 | XP_016884233.1 | 5, 6 | 793 | |||
XM_017028746.1 | XP_016884235.1 | 4, 5 | 801 | |||
XM_017028747.1 | XP_016884236.1 | 4, 5 | 801 | |||
GenBank mRNA | AB449908.1 | - | 4, 5 | 635 | ||
AK055591.1 | - | 4, 5 | 793 | |||
AK125705.1 | - | 5, 6 | 1248 | |||
AK127653.1 | - | 4, 5 | 882 | |||
AK301445.1 | - | 4, 5 | 441 | |||
AK308039.1 | - | 5, 6 | 870 | |||
AK315779.1 | - | 4, 5 | 767 | |||
BC001292.1 | - | 4, 5 | 738 | 3457393 | ||
BC020976.1 | - | 4, 5 | 740 | 3884347 | ||
BC029897.1 | - | 4, 5 | 770 | 5173045 | ||
CR456412.1 | - | 4, 5 | 656 |