Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205263.1 | NP_001192192.1 | 12 | 2640 | ||
NM_012415.3 | NP_036547.1 | 14 | 2617 | |||
GenBank mRNA | AF112481.1 | - | 14 | 2566 | ||
AK290437.1 | - | 14 | 2587 | |||
AL133578.1 | - | 8 | 1498 | |||
BC001965.2 | - | 14 | 2559 | 3452509 |