Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001234.4 | NP_001225.1 | 1 | 175 | ||
NM_033337.2 | NP_203123.1 | 1 | 175 | |||
GenBank mRNA | AF036365.1 | - | 1 | 145 | ||
AF043101.1 | - | 1 | 95 | |||
BC102033.1 | - | 1 | 135 | 40033505 | ||
BC102036.1 | - | 1 | 135 | 40033114 | ||
BC102037.1 | - | 1 | 135 | 40033118 | ||
Y14747.1 | - | 1 | 109 |